Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N-methylphthalimide 97%

Supplier: AMBEED, INC
Warning

A179899-100G A179899-10G A179899-1G A179899-250MG A179899-25G A179899-5G
76998-626EA 47 USD InStock
76998-626 76998-628 76998-630 76998-632 76998-634 76998-636
4-Amino-N-methylphthalimide 97%
4-Amino-N-methylphthalimide
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
MDL Number: MFCD00012081
CAS Number: 2307-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty