Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Heptyloxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A180738-100G A180738-25G A180738-5G
76998-664EA 155 USD InStock
76998-664 76998-666 76998-668
4-Heptyloxybenzoic acid 95%
4-Heptyloxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₀O₃
MDL Number: MFCD00013992
CAS Number: 15872-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty