Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichlorobenzophenone 98%

Supplier: AMBEED, INC
Warning

A181433-1000G A181433-100G A181433-25G A181433-500G A181433-5G
76998-678EA 194 USD InStock
76998-678 76998-680 76998-682 76998-684 76998-686 76998-688
2,5-Dichlorobenzophenone 98%
2,5-Dichlorobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₈Cl₂O
MDL Number: MFCD00079746
CAS Number: 16611-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty