Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-Benzylhydroxylamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A181641-1000G A181641-100G A181641-10G A181641-25G A181641-500G
76998-700EA 187 USD InStock
76998-700 76998-702 76998-704 76998-706 76998-708
O-Benzylhydroxylamine hydrochloride 97%
O-Benzylhydroxylamine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₀ClNO
MDL Number: MFCD00012952
CAS Number: 2687-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty