Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Phenylsulfonyl)piperidin-4-amine 95%

Supplier: AMBEED, INC
Warning

A181765-100MG A181765-1G A181765-250MG A181765-5G
76998-710EA 30 USD InStock
76998-710 76998-712 76998-714 76998-716
1-(Phenylsulfonyl)piperidin-4-amine 95%
1-(Phenylsulfonyl)piperidin-4-amine
Precaución: for Research Use Only


Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD06805738
CAS Number: 228259-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty