Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hexadecyloxy)propan-1-ol 98%

Supplier: AMBEED, INC
Warning

A182606-10G A182606-1G A182606-250MG A182606-5G
76985-788EA 275 USD InStock
76985-788 76985-790 76985-792 76985-794
3-(Hexadecyloxy)propan-1-ol 98%
3-(Hexadecyloxy)propan-1-ol
Precaución: for Research Use Only


Formula: C19H40O2
MDL Number: MFCD00171483
CAS Number: 23377-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty