Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-D-norleucine 97%

Supplier: AMBEED, INC
Warning

A183990-10G A183990-1G A183990-250MG A183990-5G A183990-100G
76998-778EA 10 USD InStock
76998-778 76998-780 76998-782 76998-784 77671-600
N-Fmoc-D-norleucine 97%
N-Fmoc-D-norleucine
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
MDL Number: MFCD00155639
CAS Number: 112883-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty