Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-fluoro-3-nitrobenzoate 98%

Supplier: AMBEED, INC
Warning

A184330-100G A184330-10G A184330-1G A184330-250MG A184330-25G A184330-5G
77029-576EA 167 USD InStock
77029-576 77029-578 77029-580 77029-582 77029-584 77029-586
Methyl-2-fluoro-3-nitrobenzoate 98%
Methyl 2-fluoro-3-nitrobenzoate
Precaución: for Research Use Only


Formula: C₈H₆FNO₄
MDL Number: MFCD10566813
CAS Number: 946126-94-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty