Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-fluoropicolinaldehyde 98%

Supplier: AMBEED, INC
Warning

A186398-100MG A186398-1G A186398-250MG A186398-25G A186398-5G
77029-642EA 5 USD InStock
77029-642 77029-644 77029-646 77029-648 77029-650
6-Chloro-3-fluoropicolinaldehyde 98%
6-Chloro-3-fluoropicolinaldehyde
Precaución: for Research Use Only


Formula: C₆H₃ClFNO
MW: 159.546 g/mol
MDL Number: MFCD04972390
CAS Number: 884494-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty