Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 95%

Supplier: AMBEED, INC
Warning

A187160-1G A187160-25G A187160-5G
77029-678EA 9 USD InStock
77029-678 77029-680 77029-682
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 95%
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₅
MW: 233.26 g/mol
MDL Number: MFCD18071557
CAS Number: 1146954-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty