Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylbenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A187266-1G A187266-25G A187266-5G
77029-684EA 6 USD InStock
77029-684 77029-686 77029-688
4-Isopropylbenzyl alcohol 98%
4-Isopropylbenzyl alcohol


Formula: C₁₀H₁₄O
MDL Number: MFCD00004663
CAS Number: 536-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty