Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dichloro-4-hydroxypyridazine 95%

Supplier: AMBEED, INC
Warning

A187308-100MG A187308-10G A187308-1G A187308-250MG A187308-5G
77029-690EA 6 USD InStock
77029-690 77029-692 77029-694 77029-696 77600-522
3,6-Dichloro-4-hydroxypyridazine 95%
3,6-Dichloro-4-hydroxypyridazine
Precaución: for Research Use Only


Formula: C₄H₂Cl₂N₂O
MW: 164.977 g/mol
MDL Number: MFCD04971276
CAS Number: 2779-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty