Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-3-methyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A188857-100MG A188857-1G A188857-250MG A188857-5G
76986-024EA 39 USD InStock
76986-024 76986-026 76986-028 76986-030
1-Boc-3-methyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Boc-3-methyl-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₅BN₂O₄
MDL Number: MFCD12407274
CAS Number: 1009071-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty