Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)benzenesulfonamide 97%

Supplier: AMBEED, INC
Danger

A189032-100MG A189032-250MG A189032-5G
77029-742EA 56 USD InStock
77029-742 77029-744 77029-746
3-(Bromomethyl)benzenesulfonamide 97%
3-(Bromomethyl)benzenesulphonamide
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂S
MDL Number: MFCD09743699
CAS Number: 220798-52-7
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty