Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorophenyl)pyridine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A189736-1G A189736-250MG
77029-772EA 254 USD InStock
77029-772 77029-774
4-(4-Fluorophenyl)pyridine-2-carboxylic acid 97%
4-(4-Fluorophenyl)pyridine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₂H₈FNO₂
MW: 217.2 g/mol
MDL Number: MFCD14666492
CAS Number: 1214388-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty