Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-3,4-dihydro-2H-isoquinolin-1-one 97%

Supplier: AMBEED, INC
Warning

A190261-100MG A190261-10G A190261-1G A190261-250MG A190261-5G
77012-278EA 13 USD InStock
77012-278 77012-280 77012-282 77012-284 77012-286
7-Bromo-3,4-dihydro-2H-isoquinolin-1-one 97%
7-Bromo-3,4-dihydro-2H-isoquinolin-1-one
Precaución: for Research Use Only


Formula: C₉H₈BrNO
MDL Number: MFCD08437642
CAS Number: 891782-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty