Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Benzyloxy)-4-iodo-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A190697-100MG A190697-1G A190697-250MG
77012-320EA 70 USD InStock
77012-320 77012-322 77012-324
1-(Benzyloxy)-4-iodo-1H-pyrazole 95%
1-(Benzyloxy)-4-iodo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₀H₉IN₂O
MW: 300.09 g/mol
MDL Number: MFCD10699161
CAS Number: 229171-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty