Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-5-bromo-2-chloropyridine 97%

Supplier: AMBEED, INC
Warning

A190994-100MG A190994-250MG
76986-108EA 21 USD InStock
76986-108 76986-110
3-(Benzyloxy)-5-bromo-2-chloropyridine 97%
3-(Benzyloxy)-5-bromo-2-chloropyridine
Precaución: for Research Use Only


Formula: C₁₂H₉BrClNO
MW: 298.56 g/mol
MDL Number: MFCD26403034
CAS Number: 891785-18-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty