Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-cyano-3-fluorobenzenecarboxylate 95%

Supplier: AMBEED, INC
Warning

A192210-1G A192210-250MG A192210-5G
76998-990EA 25 USD InStock
76998-990 76998-992 76998-994
Ethyl-4-cyano-3-fluorobenzenecarboxylate 95%
Ethyl-4-cyano-3-fluorobenzenecarboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈FNO₂
MDL Number: MFCD09607944
CAS Number: 952183-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty