Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mefloquine hydrochloride 99%

Supplier: AMBEED, INC
Warning

A192274-100MG A192274-1MG A192274-250MG A192274-25MG A192274-1G A192274-5G
77029-850EA 17 USD InStock
77029-850 77029-852 77029-854 77029-856 77667-602 77667-604
Mefloquine hydrochloride 99%
Mefloquine hydrochloride
Precaución: for Research Use Only


Formula: C₁₇H₁₆F₆N₂O
MDL Number: MFCD00797519
CAS Number: 51773-92-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty