Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-hydroxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A192836-1G A192836-250MG A192836-25G A192836-5G
76986-156EA 26 USD InStock
76986-156 76986-158 76986-160 76986-162
2-Fluoro-3-hydroxybenzaldehyde 98%
2-Fluoro-3-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅FO₂
MDL Number: MFCD11110191
CAS Number: 103438-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty