Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-fluoro-2-nitrophenol 98%

Supplier: AMBEED, INC
Danger

A193211-1G A193211-250MG A193211-25G
77029-876EA 22 USD InStock
77029-876 77029-878 77029-880
5-Chloro-4-fluoro-2-nitrophenol 98%
5-Chloro-4-fluoro-2-nitrophenol
Precaución: for Research Use Only


Formula: C6H3ClFNO3
MDL Number: MFCD18072578
CAS Number: 65001-79-8
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty