Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-(o-tolyl)-1,3,4-thiadiazole 95%

Supplier: AMBEED, INC
Warning

A193556-10G A193556-1G A193556-250MG A193556-25G A193556-5G
76999-060EA 51 USD InStock
76999-060 76999-062 76999-064 76999-066 76999-068
2-Amino-5-(o-tolyl)-1,3,4-thiadiazole 95%
2-Amino-5-(o-tolyl)-1,3,4-thiadiazole
Precaución: for Research Use Only


Formula: C₉H₉N₃S
MDL Number: MFCD00652823
CAS Number: 59565-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty