Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-2-(propylthio)pyrimidine-4,6-diol 97%

Supplier: AMBEED, INC
Warning

A194229-100MG A194229-1G A194229-250MG
76986-218EA 104 USD InStock
76986-218 76986-220 76986-222
5-Nitro-2-(propylthio)pyrimidine-4,6-diol 97%
5-Nitro-2-(propylthio)pyrimidine-4,6-diol
Precaución: for Research Use Only


Formula: C₇H₉N₃O₄S
MDL Number: MFCD26385870
CAS Number: 145783-13-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty