Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromophenyl phenyl ether (BDE No. 1) 97%

Supplier: AMBEED, INC
Warning

A194810-10G A194810-1G A194810-250MG A194810-5G
77029-934EA 133 USD InStock
77029-934 77029-936 77029-938 77029-940
2-Bromophenyl phenyl ether (BDE No. 1) 97%
2-Bromophenyl phenyl ether (BDE No. 1)
Precaución: for Research Use Only


Formula: C₁₂H₉BrO
MDL Number: MFCD00626462
CAS Number: 7025-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty