Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-2,3-dihydro-1-benzofuran 97%

Supplier: AMBEED, INC
Warning

A195308-10G A195308-1G A195308-25G A195308-5G
76986-254EA 40 USD InStock
76986-254 76986-256 76986-258 76986-260 76986-262
5-Nitro-2,3-dihydro-1-benzofuran 97%
5-Nitro-2,3-dihydro-1-benzofuran
Precaución: for Research Use Only


Formula: C₈H₇NO₃
MDL Number: MFCD22376562
CAS Number: 17403-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty