Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-1H-1,2,4-triazole-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A195366-100MG A195366-1G A195366-250MG
76986-264EA 18 USD InStock
76986-264 76986-266 76986-268
1-Benzyl-1H-1,2,4-triazole-3-carboxylic acid 97%
1-Benzyl-1H-1,2,4-triazole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₉N₃O₂
MDL Number: MFCD13191550
CAS Number: 138624-97-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty