Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-1H-benzo[d][1,2,3]triazole 95%

Supplier: AMBEED, INC
Warning

A195382-100MG A195382-1G A195382-250MG
77012-490EA 31 USD InStock
77012-490 77012-492 77012-494
1-Phenyl-1H-benzo[d][1,2,3]triazole 95%
1-Phenyl-1H-benzo[d][1,2,3]triazole
Precaución: for Research Use Only


Formula: C₁₂H₉N₃
MDL Number: MFCD01924397
CAS Number: 883-39-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty