Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Chlorophenyl)glutaramic acid 95%

Supplier: AMBEED, INC
Warning

A195633-100G A195633-10G A195633-25G A195633-5G
76999-104EA 73 USD InStock
76999-104 76999-106 76999-108 76999-110 76999-112
3-(4-Chlorophenyl)glutaramic acid 95%
3-(4-Chlorophenyl)glutaramic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD00190251
CAS Number: 1141-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty