Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chlorothiophene-2-sulfonic acid tert-butylamide 98%

Supplier: AMBEED, INC
Warning

A195656-100G A195656-10G A195656-1G A195656-25G A195656-5G
76999-116EA 112 USD InStock
76999-116 76999-118 76999-120 76999-122 76999-124
5-Chlorothiophene-2-sulfonic acid tert-butylamide 98%
5-Chlorothiophene-2-sulfonic acid tert-butylamide
Precaución: for Research Use Only


Formula: C₈H₁₂ClNO₂S₂
MW: 253.77 g/mol
MDL Number: MFCD05086586
CAS Number: 155731-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty