Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylcinnamic acid 99%

Supplier: AMBEED, INC
Warning

A196158-100G A196158-10G A196158-1G A196158-25G A196158-5G A196158-500G
76999-140EA 37 USD InStock
76999-140 76999-142 76999-144 76999-146 76999-148 77671-742
4-Methylcinnamic acid 99%
4-Methylcinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00002697
CAS Number: 1866-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty