Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A196462-100MG A196462-10G A196462-1G A196462-250MG A196462-25G A196462-5G
76999-158EA 5 USD InStock
76999-158 76999-160 76999-162 76999-164 76999-166 76999-168
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 98%
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
MDL Number: MFCD00270364
CAS Number: 263396-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty