Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3,4-Dichlorophenyl)-1-tetralone 97%

Supplier: AMBEED, INC
Warning

A197213-100G A197213-10G A197213-25G A197213-500G A197213-5G
77030-024EA 73 USD InStock
77030-024 77030-026 77030-028 77030-030 77030-032
4-(3,4-Dichlorophenyl)-1-tetralone 97%
4-(3,4-Dichlorophenyl)-1-tetralone
Precaución: for Research Use Only


Formula: C₁₆H₁₂Cl₂O
MW: 291.18 g/mol
MDL Number: MFCD02093088
CAS Number: 79560-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty