Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-N-methylpicolinamide 97%

Supplier: AMBEED, INC
Warning

A198094-100G A198094-10G A198094-25G A198094-500G
77012-570EA 39 USD InStock
77012-570 77012-572 77012-574 77012-576 77012-578
4-Chloro-N-methylpicolinamide 97%
4-Chloro-N-methylpicolinamide
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O
MW: 170.60 g/mol
MDL Number: MFCD02185921
CAS Number: 220000-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty