Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-2(1H)-quinolinone 97%

Supplier: AMBEED, INC
Warning

A198828-100MG A198828-1G A198828-250MG
76999-242EA 13 USD InStock
76999-242 76999-244 76999-246
5-Hydroxy-2(1H)-quinolinone 97%
5-Hydroxy-2(1H)-quinolinone
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MDL Number: MFCD02181132
CAS Number: 31570-97-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty