Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Cyclopropylpyridin-2-ol 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A199306-100MG A199306-250MG
76986-386EA 43 USD InStock
76986-386 77683-552
5-Cyclopropylpyridin-2-ol 97%, Ambeed.Inc
5-Cyclopropylpyridin-2-ol
Precaución: For research use only.


Formula: C₈H₉NO
MDL Number: MFCD12033218
CAS Number: 1159821-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty