Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxy-5-bromopyridine 98%

Supplier: AMBEED, INC
Warning

A199339-1G A199339-250MG A199339-25G A199339-5G
77012-592EA 15 USD InStock
77012-592 77012-594 77012-596 77012-598
3-Benzyloxy-5-bromopyridine 98%
3-Benzyloxy-5-bromopyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₀BrNO
MW: 264.12 g/mol
MDL Number: MFCD07375004
CAS Number: 130722-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty