Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-aminopyrazolo[1,5-a]pyrimidine-3-carboxylate 96%

Supplier: AMBEED, INC
Warning

A201075-100MG A201075-1G A201075-250MG A201075-5G
76986-474EA 14 USD InStock
76986-474 76986-476 76986-478 76986-480
Ethyl 2-aminopyrazolo[1,5-a]pyrimidine-3-carboxylate 96%
Ethyl 2-aminopyrazolo[1,5-a]pyrimidine-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₀N₄O₂
MDL Number: MFCD14706232
CAS Number: 1260169-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty