Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-chloro-3-(cyanomethyl)benzoate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A201084-1G A201084-250MG A201084-25G A201084-5G A201084-10G
76986-482EA 10 USD InStock
76986-482 76986-484 76986-486 76986-488 77683-562
Methyl 4-chloro-3-(cyanomethyl)benzoate 98%, Ambeed.Inc
Methyl 4-chloro-3-(cyanomethyl)benzoate
Precaución: For research use only.


Formula: C₁₀H₈ClNO₂
MDL Number: MFCD09263465
CAS Number: 872091-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty