Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,2,2-Trifluoroethyl)-1H-pyrazole-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A201170-1G A201170-250MG A201170-5G
76999-330EA 226 USD InStock
76999-330 76999-332 76999-334
1-(2,2,2-Trifluoroethyl)-1H-pyrazole-4-carboxylic acid 97%
1-(2,2,2-Trifluoroethyl)-1H-pyrazole-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₅F₃N₂O₂
MDL Number: MFCD08556229
CAS Number: 288251-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty