Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Hydroxyphenyl)-2-methoxyethanone 98%

Supplier: AMBEED, INC
Warning

A202334-1G A202334-250MG
77012-694EA 21 USD InStock
77012-694 77012-696
1-(4-Hydroxyphenyl)-2-methoxyethanone 98%
1-(4-Hydroxyphenyl)-2-methoxyethanone
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MDL Number: MFCD08461683
CAS Number: 32136-81-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty