Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazine 95%

Supplier: AMBEED, INC
Warning

A203263-100MG A203263-250MG A203263-50MG
77012-716EA 69 USD InStock
77012-716 77012-718 77012-720
2-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazine 95%
2-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazine
Precaución: for Research Use Only


Formula: C₇H₁₁N₃
MDL Number: MFCD12198501
CAS Number: 1209273-29-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty