Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 97%

Supplier: AMBEED, INC
Warning

A203546-1G A203546-250MG A203546-5G
76986-522EA 127 USD InStock
76986-522 76986-524 76986-526
4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 97%
4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole
Precaución: for Research Use Only


Formula: C₁₆H₂₁NS₂
MDL Number: MFCD26793506
CAS Number: 141029-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty