Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Hydroxymethyl)pyrrolidin-2-one 95%

Supplier: AMBEED, INC
Warning

A204561-100MG A204561-1G A204561-250MG A204561-5G
77030-252EA 5 USD InStock
77030-252 77030-254 77030-256 77030-258
5-(Hydroxymethyl)pyrrolidin-2-one 95%
5-(Hydroxymethyl)pyrrolidin-2-one
Precaución: for Research Use Only


Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD06661931
CAS Number: 62400-75-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty