Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IWR-1-endo 99+%

Supplier: AMBEED, INC
Warning

A205590-10MG A205590-1MG A205590-50MG A205590-5MG
76986-570EA 117 USD InStock
76986-570 76986-572 76986-574 76986-576
IWR-1-endo 99+%
IWR-1-endo
Precaución: for Research Use Only


Formula: C₂₅H₁₉N₃O₃
MDL Number: MFCD18086875
CAS Number: 1127442-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty