Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dibromo-2-fluorobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A205907-100MG A205907-250MG
76986-580EA 34 USD InStock
76986-580 76986-582
4,5-Dibromo-2-fluorobenzoic acid 95%
4,5-Dibromo-2-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Br₂FO₂
MDL Number: MFCD00672987
CAS Number: 289039-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty