Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-1H-indazol-6-ol 95%

Supplier: AMBEED, INC
Warning

A206740-100MG A206740-1G A206740-250MG
77012-812EA 14 USD InStock
77012-812 77012-814 77012-816
5-Methyl-1H-indazol-6-ol 95%
5-Methyl-1H-indazol-6-ol
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD11007971
CAS Number: 1082042-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty