Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-methoxybenzothiazole 95%

Supplier: AMBEED, INC
Warning

A207249-10G A207249-1G A207249-250MG A207249-5G
76999-536EA 55 USD InStock
76999-536 76999-538 76999-540 76999-542
2-Bromo-6-methoxybenzothiazole 95%
2-Bromo-6-methoxybenzothiazole
Precaución: for Research Use Only


Formula: C₈H₆BrNOS
MW: 244.12 g/mol
MDL Number: MFCD08459026
CAS Number: 2941-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty