Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ellipticine 98%

Supplier: AMBEED, INC
Danger

A209397-10MG A209397-1MG A209397-5MG
77030-456EA 83 USD InStock
77030-456 77030-458 77030-460
Ellipticine 98%
Ellipticine
Precaución: for Research Use Only


Formula: C₁₇H₁₄N₂
MDL Number: MFCD00010524
CAS Number: 519-23-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty