Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 97%

Supplier: AMBEED, INC
Warning

A210349-10G A210349-1G A210349-25G A210349-5G
76999-598EA 95 USD InStock
76999-598 76999-600 76999-602 76999-604
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 97%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Precaución: for Research Use Only


Formula: C₄₄H₃₂N₂
MDL Number: MFCD03093246
CAS Number: 123847-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty